2-(Dimethylamino)nicotinic acid

Supplier: Matrix Scientific

051966-500MG 051966-2.5G
101897-754EA 364.62 USD
101897-754 101897-756
2-(Dimethylamino)nicotinic acid
2-(Dimethylamino)nicotinic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD07379591
CAS Number: 883787-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR