2-[Methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]isonicotinic acid

Supplier: Matrix Scientific

051965-500MG 051965-2.5G
101897-750EA 461.54 USD
101897-750 101897-752
2-[Methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]isonicotinic acid
2-[Methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD13561875
CAS Number: 1219981-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR