2-[3,4-Dihydro-1(2H)-quinolinyl]isonicotinic acid

Supplier: Matrix Scientific

051963-500MG 051963-2.5G
101897-742EA 364.62 USD
101897-742 101897-744
2-[3,4-Dihydro-1(2H)-quinolinyl]isonicotinic acid
2-[3,4-Dihydro-1(2H)-quinolinyl]isonicotinic acid

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₂
MDL Number: MFCD12622023
CAS Number: 1019387-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR