2-(2,3-Dihydro-1H-indol-1-yl)isonicotinic acid

Supplier: Matrix Scientific

051962-500MG 051962-2.5G
101897-738EA 384.62 USD
101897-738 101897-740
2-(2,3-Dihydro-1H-indol-1-yl)isonicotinic acid
2-(2,3-Dihydro-1H-indol-1-yl)isonicotinic acid

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂
MDL Number: MFCD11523633
CAS Number: 1019357-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR