2-[3,4-Dihydro-2(1H)-isoquinolinyl]isonicotinic acid

Supplier: Matrix Scientific

051961-500MG 051961-2.5G
101897-734EA 166.15 USD
101897-734 101897-736
2-[3,4-Dihydro-2(1H)-isoquinolinyl]isonicotinic acid
2-[3,4-Dihydro-2(1H)-isoquinolinyl]isonicotinic acid

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₂
MDL Number: MFCD12622004
CAS Number: 1019375-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR