2-[4-(2-Hydroxyethyl)-1-piperazinyl]isonicotinic acid

Supplier: Matrix Scientific

051960-500MG 051960-2.5G
101897-730EA 364.62 USD
101897-730 101897-732
2-[4-(2-Hydroxyethyl)-1-piperazinyl]isonicotinic acid
2-[4-(2-Hydroxyethyl)-1-piperazinyl]isonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃
MDL Number: MFCD12642110
CAS Number: 1156112-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR