2-[Cyclohexyl(ethyl)amino]isonicotinic acid

Supplier: Matrix Scientific

051959-500MG 051959-2.5G
101897-726EA 364.62 USD
101897-726 101897-728
2-[Cyclohexyl(ethyl)amino]isonicotinic acid
2-[Cyclohexyl(ethyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD11523601
CAS Number: 1019466-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR