2-[Benzyl(methyl)amino]isonicotinic acid

Supplier: Matrix Scientific

051958-500MG 051958-2.5G
101897-722EA 364.62 USD
101897-722 101897-724
2-[Benzyl(methyl)amino]isonicotinic acid
2-[Benzyl(methyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD11523576
CAS Number: 77314-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR