2-(2-Ethyl-1-piperidinyl)isonicotinic acid

Supplier: Matrix Scientific

051957-500MG 051957-2.5G
101897-718EA 364.62 USD
101897-718 101897-720
2-(2-Ethyl-1-piperidinyl)isonicotinic acid
2-(2-Ethyl-1-piperidinyl)isonicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD11618680
CAS Number: 1094711-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR