2-[Cyclohexyl(methyl)amino]isonicotinic acid

Supplier: Matrix Scientific

051956-500MG 051956-2.5G
101897-714EA 364.62 USD
101897-714 101897-716
2-[Cyclohexyl(methyl)amino]isonicotinic acid
2-[Cyclohexyl(methyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD11132109
CAS Number: 1019350-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR