2-(Dipropylamino)isonicotinic acid

Supplier: Matrix Scientific

051955-500MG 051955-2.5G
101897-710EA 364.62 USD
101897-710 101897-712
2-(Dipropylamino)isonicotinic acid
2-(Dipropylamino)isonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
MDL Number: MFCD11132060
CAS Number: 597564-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR