2-(1-Azepanyl)isonicotinic acid

Supplier: Matrix Scientific

Synonyms: 2-(Azepan-1-yl)isonicotinic acid

051952-500MG 051952-2.5G
101897-698EA 364.62 USD
101897-698 101897-700
2-(1-Azepanyl)isonicotinic acid
2-(1-Azepanyl)isonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
MDL Number: MFCD04115216
CAS Number: 885277-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR