2-(4-Benzyl-1-piperazinyl)isonicotinic acid

Supplier: Matrix Scientific

051951-500MG 051951-2.5G
101897-694EA 166.15 USD
101897-694 101897-696
2-(4-Benzyl-1-piperazinyl)isonicotinic acid
2-(4-Benzyl-1-piperazinyl)isonicotinic acid

Hazard: Irritant

Formula: C₁₇H₁₉N₃O₂
MDL Number: MFCD13561874
CAS Number: 1019373-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR