2-(4-Methyl-1-piperidinyl)isonicotinic acid
Supplier: Matrix Scientific
051950-500MG
051950-2.5G
101897-690EA
384.62
USD
101897-690
101897-692
2-(4-Methyl-1-piperidinyl)isonicotinic acid
2-(4-Methyl-1-piperidinyl)isonicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD11131949 CAS Number: 1019387-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...