2-(2-Methyl-1-piperidinyl)isonicotinic acid
Supplier: Matrix Scientific
051949-500MG
051949-2.5G
101897-686EA
364.62
USD
101897-686
101897-688
2-(2-Methyl-1-piperidinyl)isonicotinic acid
2-(2-Methyl-1-piperidinyl)isonicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD11131978 CAS Number: 1019127-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...