2-[Butyl(methyl)amino]isonicotinic acid

Supplier: Matrix Scientific

051946-500MG 051946-2.5G
101897-674EA 364.62 USD
101897-674 101897-676
2-[Butyl(methyl)amino]isonicotinic acid
2-[Butyl(methyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD11132101
CAS Number: 1019349-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR