2-[(2-Hydroxyethyl)(methyl)amino]isonicotinic acid

Supplier: Matrix Scientific

051944-500MG 051944-2.5G
101897-666EA 364.62 USD
101897-666 101897-668
2-[(2-Hydroxyethyl)(methyl)amino]isonicotinic acid
2-[(2-Hydroxyethyl)(methyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
MDL Number: MFCD12771277
CAS Number: 1184491-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR