{2-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinyl}methanamine

Supplier: Matrix Scientific

051940-500MG 051940-2.5G
101897-650EA 364.62 USD
101897-650 101897-652
{2-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinyl}methanamine
{2-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinyl}methanamine

Hazard: Irritant

Formula: C₁₅H₁₇N₃
MDL Number: MFCD09732461
CAS Number: 953884-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR