[2-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinyl]methanamine
Supplier: Matrix Scientific
051939-500MG
051939-2.5G
101897-646EA
364.62
USD
101897-646
101897-648
[2-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinyl]methanamine
[2-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinyl]methanamine
Hazard: Irritant
Formula:
C₁₄H₁₅N₃ |
MDL Number:
MFCD09732143 CAS Number: 953723-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...