{2-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-pyridinyl}methanamine

Supplier: Matrix Scientific

Synonyms: (2-(3,4-Dihydroisoquinolin-2(1H)-yl)pyridin-3-yl)methanamine

051938-500MG 051938-2.5G
101897-642EA 384.62 USD
101897-642 101897-644
{2-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-pyridinyl}methanamine
{2-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-pyridinyl}methanamine

Hazard: Irritant

Formula: C₁₅H₁₇N₃
MW: 239.32 g/mol
MDL Number: MFCD09738735
CAS Number: 954257-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR