2-{4-[3-(Aminomethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol

Supplier: Matrix Scientific

051937-500MG 051937-2.5G
101897-638EA 384.62 USD
101897-638 101897-640
2-{4-[3-(Aminomethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol
2-{4-[3-(Aminomethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol

Hazard: Irritant

Formula: C₁₂H₂₀N₄O
MDL Number: MFCD12437586
CAS Number: 1156919-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR