3-(Aminomethyl)-N-cyclohexyl-N-ethyl-2-pyridinamine

Supplier: Matrix Scientific

051936-500MG 051936-2.5G
101897-634EA 364.62 USD
101897-634 101897-636
3-(Aminomethyl)-N-cyclohexyl-N-ethyl-2-pyridinamine
3-(Aminomethyl)-N-cyclohexyl-N-ethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₄H₂₃N₃
MDL Number: MFCD09730151
CAS Number: 953723-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR