N-[3-(Aminomethyl)-2-pyridinyl]-N-benzyl-N-methylamine

Supplier: Matrix Scientific

051935-500MG 051935-2.5G
101897-630EA 364.62 USD
101897-630 101897-632
N-[3-(Aminomethyl)-2-pyridinyl]-N-benzyl-N-methylamine
N-[3-(Aminomethyl)-2-pyridinyl]-N-benzyl-N-methylamine

Hazard: Irritant

Formula: C₁₄H₁₇N₃
MDL Number: MFCD09812360
CAS Number: 1016523-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR