3-(Aminomethyl)-N-cyclohexyl-N-methyl-2-pyridinamine

Supplier: Matrix Scientific

051933-500MG 051933-2.5G
101897-622EA 364.62 USD
101897-622 101897-624
3-(Aminomethyl)-N-cyclohexyl-N-methyl-2-pyridinamine
3-(Aminomethyl)-N-cyclohexyl-N-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
MDL Number: MFCD09733288
CAS Number: 953905-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR