3-(Aminomethyl)-N,N-dipropyl-2-pyridinamine

Supplier: Matrix Scientific

051932-500MG 051932-2.5G
101897-618EA 364.62 USD
101897-618 101897-620
3-(Aminomethyl)-N,N-dipropyl-2-pyridinamine
3-(Aminomethyl)-N,N-dipropyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₂₁N₃
MDL Number: MFCD09740014
CAS Number: 954585-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR