4-[3-(Aminomethyl)-2-pyridinyl]-2-piperazinone

Supplier: Matrix Scientific

051930-500MG 051930-2.5G
101897-610EA 384.62 USD
101897-610 101897-612
4-[3-(Aminomethyl)-2-pyridinyl]-2-piperazinone
4-[3-(Aminomethyl)-2-pyridinyl]-2-piperazinone

Hazard: Irritant

Formula: C₁₀H₁₄N₄O
MDL Number: MFCD09730456
CAS Number: 953894-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR