[2-(1-Azepanyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

051929-500MG 051929-2.5G
101897-606EA 364.62 USD
101897-606 101897-608
[2-(1-Azepanyl)-3-pyridinyl]methanamine
[2-(1-Azepanyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD09740461
CAS Number: 954575-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR