[2-(4-Benzyl-1-piperazinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

051928-500MG 051928-2.5G
101897-602EA 384.62 USD
101897-602 101897-604
[2-(4-Benzyl-1-piperazinyl)-3-pyridinyl]methanamine
[2-(4-Benzyl-1-piperazinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₇H₂₂N₄
MDL Number: MFCD13561872
CAS Number: 1156865-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR