N,N-Diallyl-3-(aminomethyl)-2-pyridinamine

Supplier: Matrix Scientific

051923-500MG 051923-2.5G
101897-582EA 364.62 USD
101897-582 101897-584
N,N-Diallyl-3-(aminomethyl)-2-pyridinamine
N,N-Diallyl-3-(aminomethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₇N₃
MDL Number: MFCD13561871
CAS Number: 1220028-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR