2-[[3-(Aminomethyl)-2-pyridinyl](ethyl)amino]-1-ethanol
Supplier: Matrix Scientific
051922-500MG
051922-2.5G
101897-578EA
364.62
USD
101897-578
101897-580
2-[[3-(Aminomethyl)-2-pyridinyl](ethyl)amino]-1-ethanol
2-[[3-(Aminomethyl)-2-pyridinyl](ethyl)amino]-1-ethanol
Hazard: Irritant
Formula:
C₁₀H₁₇N₃O |
MDL Number:
MFCD12772683 CAS Number: 1183103-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...