3-(Aminomethyl)-N-butyl-N-methyl-2-pyridinamine

Supplier: Matrix Scientific

051921-500MG 051921-2.5G
101897-574EA 364.62 USD
101897-574 101897-576
3-(Aminomethyl)-N-butyl-N-methyl-2-pyridinamine
3-(Aminomethyl)-N-butyl-N-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₉N₃
MDL Number: MFCD09815681
CAS Number: 1016750-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR