1-[3-(Aminomethyl)-2-pyridinyl]-3-pyrrolidinol dihydrochloride
Supplier: Matrix Scientific
051920-500MG
051920-2.5G
101897-570EA
364.62
USD
101897-570
101897-572
1-[3-(Aminomethyl)-2-pyridinyl]-3-pyrrolidinol dihydrochloride
1-[3-(Aminomethyl)-2-pyridinyl]-3-pyrrolidinol dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₇Cl₂N₃O MW: 266.17 g/mol |
MDL Number:
MFCD28506178 CAS Number: 1820604-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...