[2-(4-Morpholinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

051919-500MG 051919-2.5G
101897-566EA 384.62 USD
101897-566 101897-568
[2-(4-Morpholinyl)-3-pyridinyl]methanamine
[2-(4-Morpholinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD03086197
CAS Number: 870063-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR