2-[[3-(Aminomethyl)-2-pyridinyl](methyl)amino]-1-ethanol

Supplier: Matrix Scientific

051917-500MG 051917-2.5G
101897-558EA 364.62 USD
101897-558 101897-560
2-[[3-(Aminomethyl)-2-pyridinyl](methyl)amino]-1-ethanol
2-[[3-(Aminomethyl)-2-pyridinyl](methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₅N₃O
MDL Number: MFCD12813429
CAS Number: 1178351-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR