6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyrazinamine

Supplier: Matrix Scientific

051913-500MG 051913-2.5G
101897-542EA 364.62 USD
101897-542 101897-544
6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyrazinamine
6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O
MDL Number: MFCD13561869
CAS Number: 1219981-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR