N-Benzyl-6-chloro-N-ethyl-2-pyrazinamine

Supplier: Matrix Scientific

051912-500MG 051912-2.5G
101897-538EA 364.62 USD
101897-538 101897-540
N-Benzyl-6-chloro-N-ethyl-2-pyrazinamine
N-Benzyl-6-chloro-N-ethyl-2-pyrazinamine

Hazard: Irritant

Formula: C₁₃H₁₄ClN₃
MDL Number: MFCD13561868
CAS Number: 1219967-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR