1-(6-Chloro-2-pyrazinyl)-1,2,3,4-tetrahydroquinoline

Supplier: Matrix Scientific

051911-500MG 051911-2.5G
101897-534EA 364.62 USD
101897-534 101897-536
1-(6-Chloro-2-pyrazinyl)-1,2,3,4-tetrahydroquinoline
1-(6-Chloro-2-pyrazinyl)-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₁₃H₁₂ClN₃
MDL Number: MFCD13561867
CAS Number: 1219981-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR