2-(6-Chloro-2-pyrazinyl)-1,2,3,4-tetrahydroisoquinoline
Supplier: Matrix Scientific
051909-500MG
051909-2.5G
101897-526EA
364.62
USD
101897-526
101897-528
2-(6-Chloro-2-pyrazinyl)-1,2,3,4-tetrahydroisoquinoline
2-(6-Chloro-2-pyrazinyl)-1,2,3,4-tetrahydroisoquinoline
Hazard: Irritant
Formula:
C₁₃H₁₂ClN₃ |
MDL Number:
MFCD13561865 CAS Number: 629658-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...