2-[4-(6-Chloro-2-pyrazinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051908-500MG 051908-2.5G
101897-522EA 166.15 USD
101897-522 101897-524
2-[4-(6-Chloro-2-pyrazinyl)-1-piperazinyl]-1-ethanol
2-[4-(6-Chloro-2-pyrazinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₅ClN₄O
MDL Number: MFCD13561864
CAS Number: 1219981-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR