6-Chloro-N-cyclohexyl-N-ethyl-2-pyrazinamine

Supplier: Matrix Scientific

051907-500MG 051907-2.5G
101897-518EA 364.62 USD
101897-518 101897-520
6-Chloro-N-cyclohexyl-N-ethyl-2-pyrazinamine
6-Chloro-N-cyclohexyl-N-ethyl-2-pyrazinamine

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃
MDL Number: MFCD13561863
CAS Number: 1220030-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR