N-Benzyl-6-chloro-N-methyl-2-pyrazinamine

Supplier: Matrix Scientific

051906-500MG 051906-2.5G
101897-514EA 364.62 USD
101897-514 101897-516
N-Benzyl-6-chloro-N-methyl-2-pyrazinamine
N-Benzyl-6-chloro-N-methyl-2-pyrazinamine

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃
MDL Number: MFCD13561862
CAS Number: 629657-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR