1-(6-Chloro-2-pyrazinyl)-4-piperidinol

Supplier: Matrix Scientific

051902-500MG 051902-2.5G
101897-498EA 461.54 USD
101897-498 101897-500
1-(6-Chloro-2-pyrazinyl)-4-piperidinol
1-(6-Chloro-2-pyrazinyl)-4-piperidinol

Hazard: Irritant

Formula: C₉H₁₂ClN₃O
MDL Number: MFCD09878941
CAS Number: 913282-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR