4-(6-Chloro-2-pyrazinyl)-2-piperazinone

Supplier: Matrix Scientific

051901-500MG 051901-2.5G
101897-494EA 364.62 USD
101897-494 101897-496
4-(6-Chloro-2-pyrazinyl)-2-piperazinone
4-(6-Chloro-2-pyrazinyl)-2-piperazinone

Hazard: Irritant

Formula: C₈H₉ClN₄O
MDL Number: MFCD13561859
CAS Number: 61655-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR