1-(6-Chloro-2-pyrazinyl)azepane

Supplier: Matrix Scientific

051900-500MG 051900-2.5G
101897-490EA 364.62 USD
101897-490 101897-492
1-(6-Chloro-2-pyrazinyl)azepane
1-(6-Chloro-2-pyrazinyl)azepane

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD13561858
CAS Number: 1138220-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR