2-Chloro-6-(4-methyl-1-piperidinyl)pyrazine

Supplier: Matrix Scientific

051897-500MG 051897-2.5G
101897-478EA 364.62 USD
101897-478 101897-480
2-Chloro-6-(4-methyl-1-piperidinyl)pyrazine
2-Chloro-6-(4-methyl-1-piperidinyl)pyrazine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD10614662
CAS Number: 885704-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR