2-Chloro-6-(2-methyl-1-piperidinyl)pyrazine

Supplier: Matrix Scientific

051895-500MG 051895-2.5G
101897-470EA 364.62 USD
101897-470 101897-472
2-Chloro-6-(2-methyl-1-piperidinyl)pyrazine
2-Chloro-6-(2-methyl-1-piperidinyl)pyrazine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD13561855
CAS Number: 1219967-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR