2-[(6-Chloro-2-pyrazinyl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

051893-500MG 051893-2.5G
101897-462EA 364.62 USD
101897-462 101897-464
2-[(6-Chloro-2-pyrazinyl)(ethyl)amino]-1-ethanol
2-[(6-Chloro-2-pyrazinyl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₈H₁₂ClN₃O
MDL Number: MFCD13561853
CAS Number: 1219967-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR