N-Butyl-6-chloro-N-methyl-2-pyrazinamine

Supplier: Matrix Scientific

051892-500MG 051892-2.5G
101897-458EA 364.62 USD
101897-458 101897-460
N-Butyl-6-chloro-N-methyl-2-pyrazinamine
N-Butyl-6-chloro-N-methyl-2-pyrazinamine

Hazard: Irritant

Formula: C₉H₁₄ClN₃
MDL Number: MFCD13561852
CAS Number: 1219961-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR