1-(6-Chloro-2-pyrazinyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051891-500MG 051891-2.5G
101897-454EA 186.15 USD
101897-454 101897-456
1-(6-Chloro-2-pyrazinyl)-3-pyrrolidinol
1-(6-Chloro-2-pyrazinyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MDL Number: MFCD09879058
CAS Number: 959239-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR