2-Chloro-6-(1-piperidinyl)pyrazine

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-(piperidin-1-yl)pyrazine

051889-500MG 051889-2.5G
101897-446EA 364.62 USD
101897-446 101897-448
2-Chloro-6-(1-piperidinyl)pyrazine
2-Chloro-6-(1-piperidinyl)pyrazine

Hazard: Irritant

Formula: C₉H₁₂ClN₃
MW: 197.67 g/mol
MDL Number: MFCD09864952
CAS Number: 343856-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR